Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,3-Dichloropropionitrile, 94%

Click to view available options
Quantity:
10g
50g
2601-89-0
C3H3Cl2N
123.964
MFCD00013809
RJJDLPQZNANQDQ-UHFFFAOYSA-N
2,3-dichloropropionitrile,propanenitrile, 2,3-dichloro,alpha,beta-dichloropropionitrile,2,3-dichloropropiononitrile,propionitrile, 2,3-dichloro-6ci,7ci,8ci,acmc-1chvi,2,3-dichloropropanitrile,#
92935
2,3-dichloropropanenitrile
C(C(C#N)Cl)Cl
2601-89-0
C3H3Cl2N
123.964
MFCD00013809
RJJDLPQZNANQDQ-UHFFFAOYSA-N
2,3-dichloropropionitrile,propanenitrile, 2,3-dichloro,alpha,beta-dichloropropionitrile,2,3-dichloropropiononitrile,propionitrile, 2,3-dichloro-6ci,7ci,8ci,acmc-1chvi,2,3-dichloropropanitrile,#
92935
2,3-dichloropropanenitrile
C(C(C#N)Cl)Cl
Density 1.35
Boiling Point 62°C to 63°C (13mmHg)
Quantity 10g
UN Number UN3276
Formula Weight 123.96
Percent Purity 94%
Chemical Name or Material 2,3-Dichloropropionitrile

GHS H Statement
H300
Fatal if swallowed.

P264b-P270-P301+P310-P330-P501c

H300

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S

EINECSNumber : 220-007-4

TSCA : No

Recommended Storage : Ambient temperatures