Alfa Aesar™ 2,3-Dichloropyridine, 99%

Manufacturer: Alfa Aesar™ A1224122

Quantity 100g

 View more versions of this product

Catalog No. AAA1224122

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2402-77-9
    Molecular Formula C5H3Cl2N
    Formula Weight 147.99
    MDL Number MFCD00006229
    UN Number UN2811
    Beilstein 109811
    Chemical Name or Material 2,3-Dichloropyridine

    Specifications

    Formula Weight 147.99
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338-P309+P311-P302+P352
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Call a POISON CENTER or doctor/physician.
    IF ON SKIN: Wash with plenty of soap and water.
    CAS 2402-77-9
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    UN Number UN2811
    Beilstein 109811
    Odor Unpleasant
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H311-H302-H315-H319-H335
    Toxic in contact with skin.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Melting Point 65° to 69°C
    Quantity 100g
    Molecular Formula C5H3Cl2N
    MDL Number MFCD00006229
    RTECS Number US8050000
    Chemical Name or Material 2,3-Dichloropyridine
    Assay 99%

    GHS H Statement
    H311-H302-H315-H319-H335
    Toxic in contact with skin.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280h-P305+P351+P338-P309+P311-P302+P352
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Call a POISON CENTER or doctor/physician.
    IF ON SKIN: Wash with plenty of soap and water.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit