Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2',3'-Dideoxyguanosine, 90+%

Click to view available options
Quantity:
25mg
50mg
100mg
85326-06-3
C10H13N5O3
251.246
MFCD00057074
OCLZPNCLRLDXJC-NTSWFWBYSA-N
2',3'-dideoxyguanosine,guanosine, 2',3'-dideoxy,2-amino-9-2r,5s-5-hydroxymethyl tetrahydrofuran-2-yl-1h-purin-6 9h-one,dideoxyguanosine,2-amino-9-2r,5s-5-hydroxymethyl tetrahydrofuran-2-yl-1h-purin-6-one,deoxy-deoxy-guanosine,2,3-didioxyguanosine,2', 3'-dideoxyguanosine,2,3-dideoxyguanosine,chembl78101
64974
2-amino-9-[(2R,5S)-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
C1CC(OC1CO)N2C=NC3=C2NC(=NC3=O)N
85326-06-3
C10H13N5O3
251.246
MFCD00057074
OCLZPNCLRLDXJC-NTSWFWBYSA-N
2',3'-dideoxyguanosine,guanosine, 2',3'-dideoxy,2-amino-9-2r,5s-5-hydroxymethyl tetrahydrofuran-2-yl-1h-purin-6 9h-one,dideoxyguanosine,2-amino-9-2r,5s-5-hydroxymethyl tetrahydrofuran-2-yl-1h-purin-6-one,deoxy-deoxy-guanosine,2,3-didioxyguanosine,2', 3'-dideoxyguanosine,2,3-dideoxyguanosine,chembl78101
64974
2-amino-9-[(2R,5S)-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
C1CC(OC1CO)N2C=NC3=C2NC(=NC3=O)N
Color White
Quantity 50mg
Formula Weight 251.24
Physical Form Powder
Percent Purity ≥90%
Chemical Name or Material 2',3'-Dideoxyguanosine

TSCA : No

Recommended Storage : Store at -20°C