Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,3-Diethyl-5-methylpyrazine, 98+%

Click to view available options
Quantity:
5g
25g
100g
18138-04-0
C9H14N2
150.225
MFCD00009635
PSINWXIDJYEXLO-UHFFFAOYSA-N
pyrazine, 2,3-diethyl-5-methyl,2-methyl-5,6-diethylpyrazine,2,3-diethyl-6-methylpyrazine,5-methyl-2,3-diethylpyrazine,2,3-diethyl-5-methyl-pyrazine,fema no. 3336,hazelnut pyrazine,pubchem8621,acmc-1ca9d,ksc491g2h
28905
2,3-diethyl-5-methylpyrazine
CCC1=NC=C(N=C1CC)C
18138-04-0
C9H14N2
150.225
MFCD00009635
PSINWXIDJYEXLO-UHFFFAOYSA-N
pyrazine, 2,3-diethyl-5-methyl,2-methyl-5,6-diethylpyrazine,2,3-diethyl-6-methylpyrazine,5-methyl-2,3-diethylpyrazine,2,3-diethyl-5-methyl-pyrazine,fema no. 3336,hazelnut pyrazine,pubchem8621,acmc-1ca9d,ksc491g2h
28905
2,3-diethyl-5-methylpyrazine
CCC1=NC=C(N=C1CC)C
Boiling Point 190°C
Density 0.949
Refractive Index 1.498
Quantity 5g
Flash Point 80°C (176°F)
Formula Weight 150.23
Percent Purity ≥98%
Chemical Name or Material 2,3-Diethyl-5-methylpyrazine

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P280-P370+P378q-P501c

H227

EINECSNumber : 242-024-6

TSCA : Yes

Recommended Storage : Ambient temperatures