Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,3-Difluoro-4-ethoxybenzeneboronic acid, 98%

Click to view available options
Quantity:
1g
5g
212386-71-5
C8H9BF2O3
201.964
MFCD09258743
OBKSFBWOZSQGGC-UHFFFAOYSA-N
2,3-difluoro-4-ethoxyphenylboronic acid,4-ethoxy-2,3-difluorophenyl boronic acid,2,3-difluoro-4-ethoxybenzeneboronic acid,boronic acid, 4-ethoxy-2,3-difluorophenyl,pubchem9562,2,3-difluoro-4-ethoxyphenylboronicacid,acmc-209fik,ksc201g9b,2,3-difluoro-4-ethoxyphenylboronic acid,4-ethoxy-2,3-difluorobenzeneboronic acid
15135235
(4-ethoxy-2,3-difluorophenyl)boronic acid
B(C1=C(C(=C(C=C1)OCC)F)F)(O)O
212386-71-5
C8H9BF2O3
201.964
MFCD09258743
OBKSFBWOZSQGGC-UHFFFAOYSA-N
2,3-difluoro-4-ethoxyphenylboronic acid,4-ethoxy-2,3-difluorophenyl boronic acid,2,3-difluoro-4-ethoxybenzeneboronic acid,boronic acid, 4-ethoxy-2,3-difluorophenyl,pubchem9562,2,3-difluoro-4-ethoxyphenylboronicacid,acmc-209fik,ksc201g9b,2,3-difluoro-4-ethoxyphenylboronic acid,4-ethoxy-2,3-difluorobenzeneboronic acid
15135235
(4-ethoxy-2,3-difluorophenyl)boronic acid
B(C1=C(C(=C(C=C1)OCC)F)F)(O)O
Quantity 1g
Melting Point 151°C to 154°C
Beilstein 19662857
Formula Weight 201.96
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 2,3-Difluoro-4-ethoxybenzeneboronic acid