Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,3-Difluoro-4-methoxybenzeneboronic acid pinacol ester, 96%

Click to view available options
Quantity:
250mg
1g
1809298-92-7
2,3-Difluoro-4-methoxyphenylboronic acid pinacol ester
1809298-92-7
2,3-Difluoro-4-methoxyphenylboronic acid pinacol ester
Quantity 250mg
Formula Weight 270.08
Physical Form Solid
Percent Purity 96%
Chemical Name or Material 2,3-Difluoro-4-methoxybenzeneboronic acid pinacol ester

TSCA : No

Recommended Storage : Ambient temperatures