Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,3-Difluoro-4-methylbenzamide, 97+%

Click to view available options
Quantity:
1g
5g
261763-35-3
C8H7F2NO
171.147
MFCD01631633
JWPKXGWPXBRCLL-UHFFFAOYSA-N
acmc-1cq0b,2,3-difluoro-p-toluamide,2,3-difluoro-4-methyl-benzamide,benzamide,2,3-difluoro-4-methyl,benzamide, 2,3-difluoro-4-methyl,2,3-bis fluoranyl-4-methyl-benzamide
2774139
2,3-difluoro-4-methylbenzamide
CC1=C(C(=C(C=C1)C(=O)N)F)F
261763-35-3
C8H7F2NO
171.147
MFCD01631633
JWPKXGWPXBRCLL-UHFFFAOYSA-N
acmc-1cq0b,2,3-difluoro-p-toluamide,2,3-difluoro-4-methyl-benzamide,benzamide,2,3-difluoro-4-methyl,benzamide, 2,3-difluoro-4-methyl,2,3-bis fluoranyl-4-methyl-benzamide
2774139
2,3-difluoro-4-methylbenzamide
CC1=C(C(=C(C=C1)C(=O)N)F)F
Quantity 1g
Melting Point 158°C to 162°C
Formula Weight 171.15
Percent Purity ≥97%
Chemical Name or Material 2,3-Difluoro-4-methylbenzamide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning