Alfa Aesar™ 2,3-Difluoro-4-methylbenzylamine, 97%

Manufacturer: Alfa Aesar™ H2671503

Quantity 1g

 View more versions of this product

Catalog No. AAH2671503

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 261763-41-1
    Molecular Formula C8H9F2N
    Formula Weight 157.16
    MDL Number MFCD01631409
    UN Number UN2735
    Chemical Name or Material 2,3-Difluoro-4-methylbenzylamine

    Specifications

    Formula Weight 157.16
    Molecular Formula C8H9F2N
    MDL Number MFCD01631409
    UN Number UN2735
    Health Hazard 2 GHS H Statement
    H314
    Causes severe skin burns and eye damage.
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Boiling Point 42°C (1mmHg)
    Quantity 1g
    CAS 261763-41-1
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
    Chemical Name or Material 2,3-Difluoro-4-methylbenzylamine
    Assay 97%

    GHS H Statement
    H314
    Causes severe skin burns and eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit