Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Difluoro-4-(trifluoromethyl)pyridine, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
1215034-77-7
C6H2F5N
183.07
ZOMVJIOHXSXCIP-UHFFFAOYSA-N
2,3-difluoro-4-trifluoromethyl pyridine,alpha,alpha,alpha,2,3-pentafluoro-4-picoline
13491638
2,3-difluoro-4-(trifluoromethyl)pyridine
C1=CN=C(C(=C1C(F)(F)F)F)F
1215034-77-7
C6H2F5N
183.07
ZOMVJIOHXSXCIP-UHFFFAOYSA-N
2,3-difluoro-4-trifluoromethyl pyridine,alpha,alpha,alpha,2,3-pentafluoro-4-picoline
13491638
2,3-difluoro-4-(trifluoromethyl)pyridine
C1=CN=C(C(=C1C(F)(F)F)F)F
Quantity 1g
Packaging Glass Bottle
Boiling Point 82.0°C to 84.0°C (400.0 Torr)
Assay Percent Range 96% min. (GC)
Formula Weight 183.07
Percent Purity 97%
Chemical Name or Material 2, 3-Difluoro-4-(trifluoromethyl)pyridine

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapour.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin

GHS Signal Word: Warning