Learn More
Alfa Aesar™ 2,3-Difluoro-6-methoxybenzylamine, 97%
Manufacturer:  Alfa Aesar™ H26670MD
Quantity 250mg
 View more versions of this product
Catalog No. AAH26670MD
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
886501-77-5
173.16
UN2735
C8H9F2NO
MFCD04116022
2,3-Difluoro-6-methoxybenzylamine
Specifications
250mg
C8H9F2NO
MFCD04116022
2,3-Difluoro-6-methoxybenzylamine
886501-77-5
173.16
UN2735
97%
Safety and Handling
SDS & Safety Documents
GHS H Statement
H314
Causes severe skin burns and eye damage.
GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Danger
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.