Alfa Aesar™ 2,3-Difluoro-6-methoxyphenol, 97%

Manufacturer: Alfa Aesar™ H2645903

Quantity 1g

 View more versions of this product

Catalog No. AAH2645903

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 186306-70-7
    Molecular Formula C7H6F2O2
    Formula Weight 160.12
    MDL Number MFCD04116024
    Chemical Name or Material 2,3-Difluoro-6-methoxyphenol

    Specifications

    Formula Weight 160.12
    Health Hazard 1 Danger
    Quantity 1g
    Molecular Formula C7H6F2O2
    MDL Number MFCD04116024
    CAS 186306-70-7
    Health Hazard 2 GHS H Statement
    H318-H302-H335-H315
    Causes serious eye damage.
    Harmful if swallowed.
    May cause respiratory irritation.
    Causes skin irritation.
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF eye irritation persists: Get medical advice/attention.
    Melting Point 44° to 47°C
    Chemical Name or Material 2,3-Difluoro-6-methoxyphenol
    Assay 97%

    GHS H Statement
    H318-H302-H335-H315
    Causes serious eye damage.
    Harmful if swallowed.
    May cause respiratory irritation.
    Causes skin irritation.

    GHS P Statement
    P280-P305+P351+P338-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF eye irritation persists: Get medical advice/attention.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit