Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,3-Difluoro-6-methylpyridine, 98%

Click to view available options
Quantity:
250mg
1g
5g
1227579-04-5
C6H5F2N
129.11
MFCD16610828
HGBWAUANMSZOIJ-UHFFFAOYSA-N
2,3-difluoro-6-methyl-pyridine
70680696
2,3-difluoro-6-methylpyridine
CC1=NC(=C(C=C1)F)F
1227579-04-5
C6H5F2N
129.11
MFCD16610828
HGBWAUANMSZOIJ-UHFFFAOYSA-N
2,3-difluoro-6-methyl-pyridine
70680696
2,3-difluoro-6-methylpyridine
CC1=NC(=C(C=C1)F)F
Color Colorless to Yellow
Quantity 250mg
Formula Weight 129.11
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 2,3-Difluoro-6-methylpyridine