Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,3-Difluoroaniline, 98%

Click to view available options
Quantity:
1g
5g
4519-40-8
C6H5F2N
129.11
MFCD00010298
YCCQGFYAVUTQFK-UHFFFAOYSA-N
2,3-difluorobenzenamine,2,3-difluorophenylamine,benzenamine, 2,3-difluoro,2,3-difluoro-phenylamine,difluoraniline,pubchem1530,acmc-1aiiw,2,3-difluorobenzenamine?,2,3-difluorophenyl amine,2,3-difluoroaniline
78278
2,3-difluoroaniline
C1=CC(=C(C(=C1)F)F)N
4519-40-8
C6H5F2N
129.11
MFCD00010298
YCCQGFYAVUTQFK-UHFFFAOYSA-N
2,3-difluorobenzenamine,2,3-difluorophenylamine,benzenamine, 2,3-difluoro,2,3-difluoro-phenylamine,difluoraniline,pubchem1530,acmc-1aiiw,2,3-difluorobenzenamine?,2,3-difluorophenyl amine,2,3-difluoroaniline
78278
2,3-difluoroaniline
C1=CC(=C(C(=C1)F)F)N
Quantity 1g
Boiling Point 68°C to 69°C (12mmHg)
Flash Point 71°C (159°F)
Odor Characteristic
UN Number UN2941
Beilstein 2716500
Formula Weight 129.11
Percent Purity 98%
Chemical Name or Material 2,3-Difluoroaniline

GHS H Statement
H311-H331-H373-H227-H302-H315-H319
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280h-P305+P351+P338-P309-P310-P302+P352
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: FLUOROANILINES