Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,3-Difluoroaniline, 98%

Click to view available options
Quantity:
1g
5g
4519-40-8
C6H5F2N
129.11
MFCD00010298
YCCQGFYAVUTQFK-UHFFFAOYSA-N
2,3-difluorobenzenamine,2,3-difluorophenylamine,benzenamine, 2,3-difluoro,2,3-difluoro-phenylamine,difluoraniline,pubchem1530,acmc-1aiiw,2,3-difluorobenzenamine?,2,3-difluorophenyl amine,2,3-difluoroaniline
78278
2,3-difluoroaniline
C1=CC(=C(C(=C1)F)F)N
4519-40-8
C6H5F2N
129.11
MFCD00010298
YCCQGFYAVUTQFK-UHFFFAOYSA-N
2,3-difluorobenzenamine,2,3-difluorophenylamine,benzenamine, 2,3-difluoro,2,3-difluoro-phenylamine,difluoraniline,pubchem1530,acmc-1aiiw,2,3-difluorobenzenamine?,2,3-difluorophenyl amine,2,3-difluoroaniline
78278
2,3-difluoroaniline
C1=CC(=C(C(=C1)F)F)N
Density 1.274
Flash Point 71°C (159°F)
Refractive Index 1.514
Odor Characteristic
Boiling Point 68°C to 69°C (12mmHg)
Quantity 5g
UN Number UN2941
Beilstein 2716500
Formula Weight 129.11
Percent Purity 98%
Chemical Name or Material 2,3-Difluoroaniline
Show More Show Less

GHS H Statement
H311-H331-H373-H227-H302-H315-H319
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c

H227-H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: FLUOROANILINES

EINECSNumber : 224-847-2

TSCA : No

Recommended Storage : Ambient temperatures