Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,3-Difluoroanisole, 97+%

Click to view available options
Quantity:
1g
5g
134364-69-5
C7H6F2O
144.121
MFCD01631562
RDOGTTNFVLSBKG-UHFFFAOYSA-N
2,3-difluoroanisole,1,2-difluoro-3-methoxy-benzene,2,3-fluoroanisole,2,3-difluoro-1-methoxybenzene,benzene, 1,2-difluoro-3-methoxy,2,3-difluoroanisol,difluro-methoxybenzen,pubchem1969,2,3-difluoro anisole,acmc-1bwhl
2769344
1,2-difluoro-3-methoxybenzene
COC1=C(C(=CC=C1)F)F
134364-69-5
C7H6F2O
144.121
MFCD01631562
RDOGTTNFVLSBKG-UHFFFAOYSA-N
2,3-difluoroanisole,1,2-difluoro-3-methoxy-benzene,2,3-fluoroanisole,2,3-difluoro-1-methoxybenzene,benzene, 1,2-difluoro-3-methoxy,2,3-difluoroanisol,difluro-methoxybenzen,pubchem1969,2,3-difluoro anisole,acmc-1bwhl
2769344
1,2-difluoro-3-methoxybenzene
COC1=C(C(=CC=C1)F)F
Flash Point 62°C (144°F)
Quantity 5g
Formula Weight 144.12
Percent Purity ≥97%
Chemical Name or Material 2,3-Difluoroanisole

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning