Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Difluorobenzylamine, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
72235-51-9
C7H7F2N
143.14
MFCD00070793
OHZUCDHZOHSBPZ-UHFFFAOYSA-N
2,3-difluorobenzylamine,2,3-difluorophenyl methanamine,2,3-difluorobenzyl amine,2,3-difluorobenzyamine,benzenemethanamine, 2,3-difluoro,2,3-difluoro-benzylamine,2,3-difluorophenyl methylamine,1-2,3-difluorophenyl methanamine,pubchem7356,2,3-difluorophenyl methamine
2736941
(2,3-difluorophenyl)methanamine
C1=CC(=C(C(=C1)F)F)CN
72235-51-9
C7H7F2N
143.14
MFCD00070793
OHZUCDHZOHSBPZ-UHFFFAOYSA-N
2,3-difluorobenzylamine,2,3-difluorophenyl methanamine,2,3-difluorobenzyl amine,2,3-difluorobenzyamine,benzenemethanamine, 2,3-difluoro,2,3-difluoro-benzylamine,2,3-difluorophenyl methylamine,1-2,3-difluorophenyl methanamine,pubchem7356,2,3-difluorophenyl methamine
2736941
(2,3-difluorophenyl)methanamine
C1=CC(=C(C(=C1)F)F)CN
Boiling Point 65°C (4.0mmHg)
Flash Point 77°C
Specific Gravity 1.223
Density 1.2230g/mL
Packaging Glass bottle
Quantity 5g
Infrared Spectrum Authentic
Color Colorless to Yellow
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Formula Weight 143.14
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 2, 3-Difluorobenzylamine
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger