Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Difluorophenylacetic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
145689-41-4
C8H6F2O2
172.13
MFCD00040968
UXSQXUSJGPVOKT-UHFFFAOYSA-N
2,3-difluorophenylacetic acid,2-2,3-difluorophenyl acetic acid,2,3-difluorophenyl acetic acid,2,3-difluorophenylaceticacid,benzeneacetic acid, 2,3-difluoro,2,3-difluorobenzeneacetic acid,rarechem al bo 0630,pubchem1916,acmc-1crot
520772
2-(2,3-difluorophenyl)acetic acid
C1=CC(=C(C(=C1)F)F)CC(=O)O
145689-41-4
C8H6F2O2
172.13
MFCD00040968
UXSQXUSJGPVOKT-UHFFFAOYSA-N
2,3-difluorophenylacetic acid,2-2,3-difluorophenyl acetic acid,2,3-difluorophenyl acetic acid,2,3-difluorophenylaceticacid,benzeneacetic acid, 2,3-difluoro,2,3-difluorobenzeneacetic acid,rarechem al bo 0630,pubchem1916,acmc-1crot
520772
2-(2,3-difluorophenyl)acetic acid
C1=CC(=C(C(=C1)F)F)CC(=O)O
Packaging Glass Bottle
Color White
Quantity 1g
Melting Point 117.0°C to 119.0°C
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97% min. (HPLC)
Formula Weight 172.13
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 2, 3-Difluorophenylacetic acid
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present

GHS Signal Word: Warning