Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Difluoropyridine-4-carboxylic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
851386-31-7
C6H3F2NO2
159.09
FFGHMEKFPCGYEG-UHFFFAOYSA-N
2,3-difluoroisonicotinic acid,2,3-difluoro-4-pyridinecarboxylic acid,2,3-difluoro-4-carboxypyridine,4-pyridinecarboxylic acid, 2,3-difluoro,2,3-fifluoroisonicotinic acid,4-carboxy-2,3-difluoropyridine,2,3-difluoroisonicotinicacid,pubchem18431,ksc447o0d,5,6-difluoropyridine-4-carboxylic acid
11240637
2,3-difluoropyridine-4-carboxylic acid
C1=CN=C(C(=C1C(=O)O)F)F
851386-31-7
C6H3F2NO2
159.09
FFGHMEKFPCGYEG-UHFFFAOYSA-N
2,3-difluoroisonicotinic acid,2,3-difluoro-4-pyridinecarboxylic acid,2,3-difluoro-4-carboxypyridine,4-pyridinecarboxylic acid, 2,3-difluoro,2,3-fifluoroisonicotinic acid,4-carboxy-2,3-difluoropyridine,2,3-difluoroisonicotinicacid,pubchem18431,ksc447o0d,5,6-difluoropyridine-4-carboxylic acid
11240637
2,3-difluoropyridine-4-carboxylic acid
C1=CN=C(C(=C1C(=O)O)F)F
Melting Point 165°C to 167°C
Quantity 5g
Assay Percent Range 97%
Formula Weight 159.09
Percent Purity 97%
Chemical Name or Material 2, 3-Difluoropyridine-4-carboxylic acid

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning