Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,3-Difluorotoluene, 97%

Manufacturer:  Alfa Aesar™ B2395803

Quantity 1g

 View more versions of this product

Catalog No. AAB2395803


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

3828-49-7
128.12
UN2388
2,3-Difluorotoluene
C7H6F2
MFCD01631506
3235341

Specifications

15°C (59°F)
115° to 116°C
3828-49-7
128.12
UN2388
2,3-Difluorotoluene
Aromatic
1g
C7H6F2
MFCD01631506
3235341
97%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P280g-P243-P305+P351+P338
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves.
Take precautionary measures against static discharge.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLUOROTOLUENES

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit