Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dihydro-2-methylbenzofuran 98.0+%, TCI America™
SDP

Catalog No. p-7126776
Click to view available options
Quantity:
10 g
25 g

Chemical Identifiers

CAS 1746-11-8
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.18
MDL Number MFCD00023003
InChI Key BWCJVGMZEQDOMY-UHFFFAOYNA-N
Synonym 2-Methyl-2,3-dihydrobenzofuran
PubChem CID 101130
IUPAC Name 2-methyl-2,3-dihydro-1-benzofuran
SMILES CC1CC2=CC=CC=C2O1

Specifications

Color Yellow
Boiling Point 198°C
Quantity 10 g
Formula Weight 134.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3-Dihydro-2-methylbenzofuran
RTECSNumber DF6653950
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.