Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dihydro-6-isocyano-1,4-benzodioxine, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
174092-82-1
C9H7NO2
161.16
MFCD06200565
AAUQOWLQNFIDQL-UHFFFAOYSA-N
2,3-dihydro-6-isocyano-1,4-benzodioxine,6-isocyano-2,3-dihydro-benzo 1,4 dioxine,6-isocyano-4-oxachromane,n-methylidyne-2,3-dihydrobenzo b 1,4 dioxin-6-aminium,pubchem16170,2h,3h-benzo e 1,4-dioxin-6-isocyanide,6-isocyano-2,3-dihydro-1,4-benzodioxin,1,4-benzodioxin,2,3-dihydro-6-isocyano,6-isocyano-2,3-dihydrobenzo b 1,4 dioxine,6-isocyano-2,3-dihydro-benzo b 1,4 dioxine
2734808
6-isocyano-2,3-dihydro-1,4-benzodioxine
[C-]#[N+]C1=CC2=C(C=C1)OCCO2
174092-82-1
C9H7NO2
161.16
MFCD06200565
AAUQOWLQNFIDQL-UHFFFAOYSA-N
2,3-dihydro-6-isocyano-1,4-benzodioxine,6-isocyano-2,3-dihydro-benzo 1,4 dioxine,6-isocyano-4-oxachromane,n-methylidyne-2,3-dihydrobenzo b 1,4 dioxin-6-aminium,pubchem16170,2h,3h-benzo e 1,4-dioxin-6-isocyanide,6-isocyano-2,3-dihydro-1,4-benzodioxin,1,4-benzodioxin,2,3-dihydro-6-isocyano,6-isocyano-2,3-dihydrobenzo b 1,4 dioxine,6-isocyano-2,3-dihydro-benzo b 1,4 dioxine
2734808
6-isocyano-2,3-dihydro-1,4-benzodioxine
[C-]#[N+]C1=CC2=C(C=C1)OCCO2
Color Yellow to Brown
Quantity 1g
Melting Point 81.0°C to 87.0°C
Packaging Glass bottle
Infrared Spectrum Authentic
NMR 94% min.
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (HPLC)
Solubility Information Solubility in water: insoluble
Formula Weight 161.16
Physical Form Powder or Chunks
Percent Purity 95%
Chemical Name or Material 2, 3-Dihydro-6-isocyano-1, 4-benzodioxine
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning