Alfa Aesar™ 2,3-Dihydrobenzo[b]furan, 98%

Manufacturer: Alfa Aesar™ A1136814

Quantity 25g

 View more versions of this product

Catalog No. AAA1136814

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 496-16-2
    Molecular Formula C8H8O
    Formula Weight 120.15
    MDL Number MFCD00005855
    Beilstein 111928
    Merck Index 14,2559
    Synonym Coumaran; 2,3-Dihydro-2,3-benzofuran
    Chemical Name or Material 2,3-Dihydrobenzo[b]furan

    Specifications

    Formula Weight 120.15
    Health Hazard 3 GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C8H8O
    MDL Number MFCD00005855
    CAS 496-16-2
    Beilstein 111928
    Merck Index 14,2559
    Synonym Coumaran; 2,3-Dihydro-2,3-benzofuran
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H227
    Combustible liquid.
    Quantity 25g
    TSCA TSCA
    Melting Point −21°C
    Boiling Point 188° to 189°C
    Flash Point 66°C (150°F)
    Chemical Name or Material 2,3-Dihydrobenzo[b]furan
    Assay 98%

    GHS H Statement
    H227
    Combustible liquid.

    GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit