Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dihydrofuran, 98+%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Glass bottle
1191-99-7
C4H6O
70.09
MFCD00003205
JKTCBAGSMQIFNL-UHFFFAOYSA-N
4,5-dihydrofuran,furan, 2,3-dihydro,furan, dihydro,dihydrofuran,2,3 dihydrofuran,dihydrofurane,2,3dihydrofuran,furan,3-dihydro,2,3-dihydrofurane,2,3-dihydro-furan
70934
CHEBI:51662
2,3-dihydrofuran
C1COC=C1
1191-99-7
C4H6O
70.09
MFCD00003205
JKTCBAGSMQIFNL-UHFFFAOYSA-N
4,5-dihydrofuran,furan, 2,3-dihydro,furan, dihydro,dihydrofuran,2,3 dihydrofuran,dihydrofurane,2,3dihydrofuran,furan,3-dihydro,2,3-dihydrofurane,2,3-dihydro-furan
70934
CHEBI:51662
2,3-dihydrofuran
C1COC=C1
Boiling Point 54°C to 55°C
Specific Gravity 0.927
Flash Point -24°C
Refractive Index 1.4220 to 1.424
Quantity 100g
Color Colorless
Density 0.9270g/mL
Packaging Glass bottle
Infrared Spectrum Authentic
Water 0.3% max.
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98% min. (GC)
Beilstein 17, III, 141
Fieser 12,189; 15,95
Solubility Information Solubility in water: slightly soluble
Formula Weight 70.09
Physical Form Liquid
Percent Purity 98+%
Chemical Name or Material 2, 3-Dihydrofuran, 98+%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Highly flammable liquid and vapor.
May form explosive peroxides.

GHS P Statement:
Keep container tightly closed.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 214-747-7

TSCA : TSCA