Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dihydropyrido[2,3-d]oxazol-2-one, 97%, Thermo Scientific™

Catalog Number AAH6623414
Click to view available options
Quantity:
25 g
100 g
60832-72-6
C6H4N2O2
136.11
MFCD00204215,MFCD09065038
OVLXOTUWFLHWQT-UHFFFAOYSA-N
oxazolo 4,5-b pyridin-2 3h-one, 3h-oxazolo 4,5-b pyridin-2-one, 2,3-dihydropyrido 2,3-d 1,3 oxazol-2-one, 1,3 oxazolo 4,5-b pyridin-2-ol, 1,3 oxazolo 4,5-b pyridin-2 3h-one, 3h-1,3 oxazolo 4,5-b pyridin-2-one, 2h,3h-1,3 oxazolo 4,5-b pyridin-2-one, pubchem11061, 2-oxooxazolo 4,5-b pyridine
2799900
CHEBI:38580
2H,3H-[1,3]oxazolo[4,5-b]pyridin-2-one
O=C1NC2=NC=CC=C2O1
60832-72-6
C6H4N2O2
136.11
MFCD00204215,MFCD09065038
OVLXOTUWFLHWQT-UHFFFAOYSA-N
oxazolo 4,5-b pyridin-2 3h-one, 3h-oxazolo 4,5-b pyridin-2-one, 2,3-dihydropyrido 2,3-d 1,3 oxazol-2-one, 1,3 oxazolo 4,5-b pyridin-2-ol, 1,3 oxazolo 4,5-b pyridin-2 3h-one, 3h-1,3 oxazolo 4,5-b pyridin-2-one, 2h,3h-1,3 oxazolo 4,5-b pyridin-2-one, pubchem11061, 2-oxooxazolo 4,5-b pyridine
2799900
CHEBI:38580
2H,3H-[1,3]oxazolo[4,5-b]pyridin-2-one
O=C1NC2=NC=CC=C2O1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 25 g
Beilstein 125648
Formula Weight 136.11
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2,3-Dihydropyrido[2,3-d]oxazol-2-one
Hazard Category H302+H312+H332-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit