Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dihydroxybenzaldehyde, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
24677-78-9
C7H6O3
138.12
MFCD00003324
IXWOUPGDGMCKGT-UHFFFAOYSA-N
o-pyrocatechualdehyde,5,6-dihydroxybenzaldehyde,benzaldehyde, 2,3-dihydroxy,benzaldehyde, dihydroxy,2,3-dihydroxybenzaldehyde,dihydroxybenzaldehyde,pubchem6561,acmc-1clux,2,3-dihydroxy benzaldehyde,2,3-dihydroxy-benzaldehyde
90579
CHEBI:50197
2,3-dihydroxybenzaldehyde
C1=CC(=C(C(=C1)O)O)C=O
24677-78-9
C7H6O3
138.12
MFCD00003324
IXWOUPGDGMCKGT-UHFFFAOYSA-N
o-pyrocatechualdehyde,5,6-dihydroxybenzaldehyde,benzaldehyde, 2,3-dihydroxy,benzaldehyde, dihydroxy,2,3-dihydroxybenzaldehyde,dihydroxybenzaldehyde,pubchem6561,acmc-1clux,2,3-dihydroxy benzaldehyde,2,3-dihydroxy-benzaldehyde
90579
CHEBI:50197
2,3-dihydroxybenzaldehyde
C1=CC(=C(C(=C1)O)O)C=O
Quantity 25g
Packaging Glass bottle
Color Beige-Green to Yellow
Infrared Spectrum Authentic
Melting Point 104.0°C to 109.0°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula (HO)2C6H3CHO
Beilstein 08, 240
Formula Weight 138.12
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 2, 3-Dihydroxybenzaldehyde, 97%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 246-398-1

TSCA : TSCA