Alfa Aesar™ 2,3-Dihydroxynaphthalene, 98%

Manufacturer: Alfa Aesar™ A1130730

Quantity 250g

 View more versions of this product

Catalog No. AAA1130730

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 92-44-4
    Molecular Formula C10H8O2
    Formula Weight 160.17
    MDL Number MFCD00004073
    Beilstein 742375
    Synonym 2,3-Naphthalenediol
    Chemical Name or Material 2,3-Dihydroxynaphthalene

    Specifications

    Formula Weight 160.17
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C10H8O2
    MDL Number MFCD00004073
    Beilstein 742375
    Health Hazard 2 GHS H Statement
    H301-H318-H315-H335
    Toxic if swallowed.
    Causes serious eye damage.
    Causes skin irritation.
    May cause respiratory irritation.
    RTECS Number QJ4750000
    Melting Point 162° to 165°C
    TSCA TSCA
    Health Hazard 1 Danger
    Flash Point 175°C (347°F)
    CAS 92-44-4
    Synonym 2,3-Naphthalenediol
    Quantity 250g
    Chemical Name or Material 2,3-Dihydroxynaphthalene
    Assay 98%

    Description

    Applications
    2,3-Dihydroxynaphthalene is used in making dyes, pigments, fluorescent whiteners, tanning agents, antioxidants, and antiseptics.

    Solubility
    Slightly soluble in water

    Notes
    Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

    GHS H Statement
    H301-H318-H315-H335
    Toxic if swallowed.
    Causes serious eye damage.
    Causes skin irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit