Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimethoxy-5-methyl-1,4-benzoquinone, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
605-94-7
C9H10O4
182.18
UIXPTCZPFCVOQF-UHFFFAOYSA-N
2,3-dimethoxy-5-methyl-1,4-benzoquinone,coenzyme q0,ubiquinone 0,2,3-dimethoxy-5-methyl-p-benzoquinone,ubiquinone q0,ubiquinone-o,ubiquinone-0,coq0,2-methyl-4,5-dimethoxy-p-quinone,2,3-dimethoxy-5-methylbenzoquinone
69068
CHEBI:27906
2,3-dimethoxy-5-methylcyclohexa-2,5-diene-1,4-dione
CC1=CC(=O)C(=C(C1=O)OC)OC
605-94-7
C9H10O4
182.18
UIXPTCZPFCVOQF-UHFFFAOYSA-N
2,3-dimethoxy-5-methyl-1,4-benzoquinone,coenzyme q0,ubiquinone 0,2,3-dimethoxy-5-methyl-p-benzoquinone,ubiquinone q0,ubiquinone-o,ubiquinone-0,coq0,2-methyl-4,5-dimethoxy-p-quinone,2,3-dimethoxy-5-methylbenzoquinone
69068
CHEBI:27906
2,3-dimethoxy-5-methylcyclohexa-2,5-diene-1,4-dione
CC1=CC(=O)C(=C(C1=O)OC)OC
Melting Point 59°C to 61°C
Packaging Glass Bottle
Quantity 1g
Color Red to Orange
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 182.18
Physical Form Crystalline Powder or Needles
Percent Purity 98%
Chemical Name or Material 2, 3-Dimethoxy-5-methyl-1, 4-benzoquinone
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 210-100-8