Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimethoxybenzaldehyde, 97%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
86-51-1
C9H10O3
166.18
MFCD00003309
JIVGSHFYXPRRSZ-UHFFFAOYSA-N
o-veratraldehyde,benzaldehyde, 2,3-dimethoxy,unii-8alp3sy00l,2,3-dimethoxy-benzaldehyde,8alp3sy00l,benzaldehyde,3-dimethoxy,dimethoxybenzaldehyde,pubchem8216,benzaldehyde, dimethoxy,upcmld00wstructure66
66581
2,3-dimethoxybenzaldehyde
COC1=CC=CC(=C1OC)C=O
86-51-1
C9H10O3
166.18
MFCD00003309
JIVGSHFYXPRRSZ-UHFFFAOYSA-N
o-veratraldehyde,benzaldehyde, 2,3-dimethoxy,unii-8alp3sy00l,2,3-dimethoxy-benzaldehyde,8alp3sy00l,benzaldehyde,3-dimethoxy,dimethoxybenzaldehyde,pubchem8216,benzaldehyde, dimethoxy,upcmld00wstructure66
66581
2,3-dimethoxybenzaldehyde
COC1=CC=CC(=C1OC)C=O
Flash Point >110°C
Melting Point 49°C to 54°C
Boiling Point 137°C (12.0mmHg)
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 50g
Color White to Beige
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula (CH3O)2C6H3CHO
Beilstein 08, I, 601
Formula Weight 166.18
Physical Form Crystalline Powder, Crystals, and/or Chunks
Percent Purity ≥96.0%
Chemical Name or Material 2, 3-Dimethoxybenzaldehyde, 97%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 201-677-7

TSCA : TSCA