Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimethoxybenzaldehyde, 97%, Thermo Scientific Chemicals

Catalog Number p-114181
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
86-51-1
C9H10O3
166.18
MFCD00003309
JIVGSHFYXPRRSZ-UHFFFAOYSA-N
o-veratraldehyde, benzaldehyde, 2,3-dimethoxy, unii-8alp3sy00l, 2,3-dimethoxy-benzaldehyde, 8alp3sy00l, benzaldehyde,3-dimethoxy, dimethoxybenzaldehyde, pubchem8216, benzaldehyde, dimethoxy, upcmld00wstructure66
66581
2,3-dimethoxybenzaldehyde
COC1=CC=CC(=C1OC)C=O
86-51-1
C9H10O3
166.18
MFCD00003309
JIVGSHFYXPRRSZ-UHFFFAOYSA-N
o-veratraldehyde, benzaldehyde, 2,3-dimethoxy, unii-8alp3sy00l, 2,3-dimethoxy-benzaldehyde, 8alp3sy00l, benzaldehyde,3-dimethoxy, dimethoxybenzaldehyde, pubchem8216, benzaldehyde, dimethoxy, upcmld00wstructure66
66581
2,3-dimethoxybenzaldehyde
COC1=CC=CC(=C1OC)C=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 49°C to 54°C
Color White to Beige
Boiling Point 137°C (12.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (CH3O)2C6H3CHO
Quantity 10 g
Beilstein 08, I, 601
Formula Weight 166.18
Percent Purity 97%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material 2, 3-Dimethoxybenzaldehyde, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/
EINECSNumber 201-677-7
TSCA TSCA

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit