Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,3-Dimethoxybenzeneboronic acid, 98%

Click to view available options
Quantity:
1g
5g
25g
40972-86-9
C8H11BO4
181.982
MFCD02683112
VREWSCMOGIXMDQ-UHFFFAOYSA-N
2,3-dimethoxybenzeneboronic acid,2,3-dimethoxyphenyl boronic acid,boronic acid, 2,3-dimethoxyphenyl,2,3-dimethoxyphenylboronicacid,3-boronoveratrole,pubchem14109,acmc-209jhv,ksc235o1n,2,3-dimethoxy-phenylboronic acid
5156491
(2,3-dimethoxyphenyl)boronic acid
B(C1=C(C(=CC=C1)OC)OC)(O)O
40972-86-9
C8H11BO4
181.982
MFCD02683112
VREWSCMOGIXMDQ-UHFFFAOYSA-N
2,3-dimethoxybenzeneboronic acid,2,3-dimethoxyphenyl boronic acid,boronic acid, 2,3-dimethoxyphenyl,2,3-dimethoxyphenylboronicacid,3-boronoveratrole,pubchem14109,acmc-209jhv,ksc235o1n,2,3-dimethoxy-phenylboronic acid
5156491
(2,3-dimethoxyphenyl)boronic acid
B(C1=C(C(=CC=C1)OC)OC)(O)O
Melting Point 67°C to 71°C
Quantity 1g
Beilstein 3051295
Formula Weight 181.98
Percent Purity 98%
Chemical Name or Material 2,3-Dimethoxybenzeneboronic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning