Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimethoxybenzoic acid, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
1521-38-6
C9H10O4
182.18
MFCD00002432
FODBVCSYJKNBLO-UHFFFAOYSA-N
o-veratric acid,benzoic acid, 2,3-dimethoxy,benzoic acid, dimethoxy,2,3-dimethoxy benzoic acid,dimethoxybenzoic acid,2,3,o-veratric acid 6ci,7ci,8ci,2,3-dimethoxybenzoicacid,dimethoxybenzoic acid,pubchem4948,2,3-dimethoxy-benzoicaci
15204
2,3-dimethoxybenzoic acid
COC1=CC=CC(=C1OC)C(=O)O
1521-38-6
C9H10O4
182.18
MFCD00002432
FODBVCSYJKNBLO-UHFFFAOYSA-N
o-veratric acid,benzoic acid, 2,3-dimethoxy,benzoic acid, dimethoxy,2,3-dimethoxy benzoic acid,dimethoxybenzoic acid,2,3,o-veratric acid 6ci,7ci,8ci,2,3-dimethoxybenzoicacid,dimethoxybenzoic acid,pubchem4948,2,3-dimethoxy-benzoicaci
15204
2,3-dimethoxybenzoic acid
COC1=CC=CC(=C1OC)C(=O)O
Quantity 5g
Color Beige
Infrared Spectrum Authentic
Melting Point 121°C to 124°C
Packaging Glass bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula (CH3O)2C6H3CO2H
Beilstein 10, I, 174
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ethanol
Formula Weight 182.18
Physical Form Fine Crystalline Powder
Percent Purity ≥98.5%
Chemical Name or Material 2, 3-Dimethoxybenzoic acid
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.