Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimethoxyphenethylamine, 97%, Acros Organics

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
3213-29-4
C10H15NO2
181.235
XKBUFTXNLBWTFP-UHFFFAOYSA-N
2,3-dimethoxyphenethylamine,2-2,3-dimethoxyphenyl ethanamine,benzeneethanamine, 2,3-dimethoxy,2-2,3-dimethoxyphenyl ethan-1-amine,benzeneethanamine,2,3-dimethoxy,pubchem12526,dimethoxybenzeneethan-amine,acmc-1aez4,2,3-dimethoxybenzeneethanamine,2,3-dimethoxy-benzyl methylamine
137858
2-(2,3-dimethoxyphenyl)ethanamine
COC1=CC=CC(=C1OC)CCN
3213-29-4
C10H15NO2
181.235
XKBUFTXNLBWTFP-UHFFFAOYSA-N
2,3-dimethoxyphenethylamine,2-2,3-dimethoxyphenyl ethanamine,benzeneethanamine, 2,3-dimethoxy,2-2,3-dimethoxyphenyl ethan-1-amine,benzeneethanamine,2,3-dimethoxy,pubchem12526,dimethoxybenzeneethan-amine,acmc-1aez4,2,3-dimethoxybenzeneethanamine,2,3-dimethoxy-benzyl methylamine
137858
2-(2,3-dimethoxyphenyl)ethanamine
COC1=CC=CC(=C1OC)CCN
Quantity 5g
Packaging Glass bottle
Assay Percent Range 97%
Formula Weight 181.23
Chemical Name or Material 2, 3-Dimethoxyphenethylamine