Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,3-Dimethoxytoluene, 98+%

Manufacturer:  Alfa Aesar™ A1161706

Quantity 5g

 View more versions of this product

Catalog No. AAA1161706


  Check Availability
Add to cart

Specifications

Description

Applications
2,3-Dimethoxytoluene has been used to prepare 6,7-dimethoxy-4-hydroxyisochroman-3-one.

Solubility
Soluble in Dichloromethane. Not miscible in water.

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
Description & Specifications

Chemical Identifiers

4463-33-6
152.19
1940962
2,3-Dimethoxytoluene
C9H12O2
MFCD00008379
3-Methylveratrole

Specifications

85°C (185°F)
202° to 203°C
4463-33-6
152.19
1940962
2,3-Dimethoxytoluene
Aromatic
5g
C9H12O2
MFCD00008379
3-Methylveratrole
≥98%

Safety and Handling

SDS & Safety Documents

Warning

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit