Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimethyl-1-(4-methylphenyl)-3-pyrazolin-5-one, ≥99%, Acros Organics

Click to view available options
Quantity:
25mg
Packaging:
Glass bottle
56430-08-1
C12H14N2O
202.257
MFCD00003140
CSVSLJZHBKTVNT-UHFFFAOYSA-N
1,5-dimethyl-2-p-tolyl-1h-pyrazol-3 2h-one,2,3-dimethyl-1-4-methylphenyl-3-pyrazolin-5-one,1,2-dihydro-1,5-dimethyl-2-p-tolyl-3h-pyrazol-3-one,1,5-dimethyl-2-4-methylphenyl pyrazol-3-one,4-methylphenazone,1,5-dimethyl-2-4-methylphenyl-1,2-dihydro-3h-pyrazol-3-one,4 inverted exclamation marka-methylphenazone,4 inverted exclamation mark-methylphenazone,2?3-dimethyl-1-4-methylphenyl-3-pyrazolin-5-one,1,5-dimethyl-2-4-methylphenyl-2,3-dihydro-1h-pyrazol-3-one
91854
1,5-dimethyl-2-(4-methylphenyl)pyrazol-3-one
CC1=CC=C(C=C1)N2C(=O)C=C(N2C)C
56430-08-1
C12H14N2O
202.257
MFCD00003140
CSVSLJZHBKTVNT-UHFFFAOYSA-N
1,5-dimethyl-2-p-tolyl-1h-pyrazol-3 2h-one,2,3-dimethyl-1-4-methylphenyl-3-pyrazolin-5-one,1,2-dihydro-1,5-dimethyl-2-p-tolyl-3h-pyrazol-3-one,1,5-dimethyl-2-4-methylphenyl pyrazol-3-one,4-methylphenazone,1,5-dimethyl-2-4-methylphenyl-1,2-dihydro-3h-pyrazol-3-one,4 inverted exclamation marka-methylphenazone,4 inverted exclamation mark-methylphenazone,2?3-dimethyl-1-4-methylphenyl-3-pyrazolin-5-one,1,5-dimethyl-2-4-methylphenyl-2,3-dihydro-1h-pyrazol-3-one
91854
1,5-dimethyl-2-(4-methylphenyl)pyrazol-3-one
CC1=CC=C(C=C1)N2C(=O)C=C(N2C)C
Quantity 25mg
Color Brown to White
Melting Point 130°C to 137°C
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Beilstein 24, 39
Formula Weight 202.26
Physical Form Powder
Percent Purity ≥99.0%
Chemical Name or Material 2, 3-Dimethyl-1-(4-methylphenyl)-3-pyrazolin-5-one, 99+%
Show More Show Less