Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimethyl-2,3-diphenylbutane, 95%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
1889-67-4
C18H22
238.37
MFCD00053713
HGTUJZTUQFXBIH-UHFFFAOYSA-N
2,3-dimethyl-2,3-diphenylbutane,dicumene,2,3-dimethylbutane-2,3-diyl dibenzene,dicumyl,butane, 2,3-dimethyl-2,3-diphenyl,benzene, 1,1'-1,1,2,2-tetramethyl-1,2-ethanediyl bis,.alpha.,.alpha.'-dicumyl,2,3-dimethyl-3-phenylbutan-2-yl benzene,bibenzyl, .alpha.,.alpha.,.alpha.',.alpha.'-tetramethyl,1,1,2-trimethyl-2-phenylpropyl benzene
74681
(2,3-dimethyl-3-phenylbutan-2-yl)benzene
CC(C)(C1=CC=CC=C1)C(C)(C)C2=CC=CC=C2
1889-67-4
C18H22
238.37
MFCD00053713
HGTUJZTUQFXBIH-UHFFFAOYSA-N
2,3-dimethyl-2,3-diphenylbutane,dicumene,2,3-dimethylbutane-2,3-diyl dibenzene,dicumyl,butane, 2,3-dimethyl-2,3-diphenyl,benzene, 1,1'-1,1,2,2-tetramethyl-1,2-ethanediyl bis,.alpha.,.alpha.'-dicumyl,2,3-dimethyl-3-phenylbutan-2-yl benzene,bibenzyl, .alpha.,.alpha.,.alpha.',.alpha.'-tetramethyl,1,1,2-trimethyl-2-phenylpropyl benzene
74681
(2,3-dimethyl-3-phenylbutan-2-yl)benzene
CC(C)(C1=CC=CC=C1)C(C)(C)C2=CC=CC=C2
Melting Point 90°C to 110°C
Color White to Yellow
Packaging Glass bottle
Quantity 25g
Infrared Spectrum Authentic
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min
Linear Formula C6H5C(CH3)2C(CH3)2C6H5
Beilstein 05,396
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in toluene
Formula Weight 238.37
Physical Form Flakes, Crystalline Powder or Chunks
Percent Purity 95%
Chemical Name or Material 2, 3-Dimethyl-2, 3-diphenylbutane, 95%
Show More Show Less

EINECSNumber : 217-568-2

TSCA : TSCA