Alfa Aesar™ 2,3-Dimethyl-3-hexanol, 95%

Manufacturer: Alfa Aesar™ B2452514

Quantity 25g

 View more versions of this product

Catalog No. AAB2452514

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 4166-46-5
    Molecular Formula C8H18O
    Formula Weight 130.23
    MDL Number MFCD00046594
    UN Number UN1993
    Beilstein 1732656
    Synonym Isopropyl methyl n-propyl carbinol
    Chemical Name or Material 2,3-Dimethyl-3-hexanol

    Specifications

    Formula Weight 130.23
    MDL Number MFCD00046594
    CAS 4166-46-5
    UN Number UN1993
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H226
    Flammable liquid and vapour.
    Flash Point 49°C (120°F)
    Boiling Point 157° to 159°C
    Molecular Formula C8H18O
    Beilstein 1732656
    Synonym Isopropyl methyl n-propyl carbinol
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Quantity 25g
    Chemical Name or Material 2,3-Dimethyl-3-hexanol
    Assay 95%

    GHS H Statement
    H226
    Flammable liquid and vapour.

    GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit