Alfa Aesar™ 2,3-Dimethyl-6-nitroaniline, 98%

Manufacturer: Alfa Aesar™ A10412-18

Quantity 50g

 View more versions of this product

Catalog No. AAA1041218

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 59146-96-2
    Molecular Formula C8H10N2O2
    Formula Weight 166.18
    MDL Number MFCD00010148
    UN Number UN2811
    Synonym 6-Nitro-2,3-xylidine
    Chemical Name or Material 2,3-Dimethyl-6-nitroaniline

    Specifications

    Formula Weight 166.18
    Molecular Formula C8H10N2O2
    Melting Point 115° to 118°C
    MDL Number MFCD00010148
    CAS 59146-96-2
    UN Number UN2811
    Synonym 6-Nitro-2,3-xylidine
    Health Hazard 2 GHS H Statement
    H331-H302-H312-H315-H319-H335
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Quantity 50g
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for sev
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    Health Hazard 1 Danger
    Chemical Name or Material 2,3-Dimethyl-6-nitroaniline
    Assay 98%

    GHS H Statement
    H331-H302-H312-H315-H319-H335
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for sev

    Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit