Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimethylaniline, 99%, ACROS Organics™

Click to view available options
Quantity:
5mL
500mL
Packaging:
Glass bottle
87-59-2
C8H11N
121.183
MFCD00007732
VVAKEQGKZNKUSU-UHFFFAOYSA-N
2,3-xylidine,3-amino-o-xylene,benzenamine, 2,3-dimethyl,2,3-dimethylbenzenamine,2,3-dimethylphenylamine,2,3-xylylamine,ortho-xylidine,vic-o-xylidine,unii-zd450abi9x,ccris 4739
6893
2,3-dimethylaniline
CC1=C(C(=CC=C1)N)C
87-59-2
C8H11N
121.183
MFCD00007732
VVAKEQGKZNKUSU-UHFFFAOYSA-N
2,3-xylidine,3-amino-o-xylene,benzenamine, 2,3-dimethyl,2,3-dimethylbenzenamine,2,3-dimethylphenylamine,2,3-xylylamine,ortho-xylidine,vic-o-xylidine,unii-zd450abi9x,ccris 4739
6893
2,3-dimethylaniline
CC1=C(C(=CC=C1)N)C
Density 0.9900g/mL
Quantity 500mL
Refractive Index 1.5670 to 1.5690 (20°C, 589nm)
Infrared Spectrum Authentic
Packaging Glass bottle
Color Undesignated
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula (CH3)2C6H3NH2
Beilstein 12, 1101
Merck Index 15, 10282
Formula Weight 121.18
Physical Form Liquid
Percent Purity 98.5 to 101.5% total base, ≥98.5% (GC)
Chemical Name or Material 2, 3-Dimethylaniline, 99%
Show More Show Less

GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Toxic to aquatic life with long lasting effects.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 201-755-0

RTECSNumber : ZE8750000

TSCA : TSCA

Recommended Storage : product darkens if exposed to light and air