Learn More
2,3-Dimethylanisole, 97%, Thermo Scientific Chemicals
Description
2,3-Dimethylanisole is used as starting reagent in the synthesis of biphenyl-indanone A, a positive allosteric modulator of the metabotropic glutamate receptor subtype 2. It can be used to produce 2-methoxy-6-methyl-benzaldehyde by heating.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2,3-Dimethylanisole is used as starting reagent in the synthesis of biphenyl-indanone A, a positive allosteric modulator of the metabotropic glutamate receptor subtype 2. It can be used to produce 2-methoxy-6-methyl-benzaldehyde by heating.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Melting Point | 29°C |
Density | 0.984 |
Boiling Point | 195°C |
Quantity | 10 g |
Refractive Index | 1.52 |
Beilstein | 1859736 |
Solubility Information | Not miscible or difficult to mix in water. |
Formula Weight | 136.19 |
Percent Purity | 97% |
Chemical Name or Material | 2,3-Dimethylanisole |
Safety and Handling
Hazard Category | H227-H315-H319 |
Hazard Statement | GHS H Statement H227 Combustible liquid. |
Precautionary Statement | P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c |
EINECSNumber | 220-948-0 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.