Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,3-Dimethylbenzeneboronic acid, 98%

Click to view available options
Quantity:
1g
5g
25g
183158-34-1
C8H11BO2
149.984
MFCD01863524
ZYYANAWVBDFAHY-UHFFFAOYSA-N
2,3-dimethylbenzeneboronic acid,2,3-dimethylphenyl boronic acid,o-xylene-3-boronic acid,2,3-dimethylphenyl boranediol,2,3-dimethylbenzeneboronicacid,2,3-dimethylphenylboronicacid,pubchem5037,acmc-1bxk2,amtb394,ksc174k9l
2773395
(2,3-dimethylphenyl)boronic acid
B(C1=C(C(=CC=C1)C)C)(O)O
183158-34-1
C8H11BO2
149.984
MFCD01863524
ZYYANAWVBDFAHY-UHFFFAOYSA-N
2,3-dimethylbenzeneboronic acid,2,3-dimethylphenyl boronic acid,o-xylene-3-boronic acid,2,3-dimethylphenyl boranediol,2,3-dimethylbenzeneboronicacid,2,3-dimethylphenylboronicacid,pubchem5037,acmc-1bxk2,amtb394,ksc174k9l
2773395
(2,3-dimethylphenyl)boronic acid
B(C1=C(C(=CC=C1)C)C)(O)O
Quantity 1g
Melting Point ~175°C
Beilstein 8050763
Formula Weight 149.99
Percent Purity 98%
Chemical Name or Material 2,3-Dimethylbenzeneboronic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning