Alfa Aesar™ 2,3-Dimethylbutane, 99%

Manufacturer: Alfa Aesar™ B2222306

Quantity 5g

 View more versions of this product

Catalog No. AAB2222306

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 79-29-8
    Molecular Formula C6H14
    Formula Weight 86.18
    MDL Number MFCD00008925
    UN Number UN2457
    Beilstein 1730737
    Synonym Diisopropyl
    Chemical Name or Material 2,3-Dimethylbutane

    Specifications

    Formula Weight 86.18
    RTECS Number EJ9350000
    Odor Mild
    Melting Point −129°C
    Quantity 5g
    TSCA TSCA
    Molecular Formula C6H14
    MDL Number MFCD00008925
    UN Number UN2457
    Beilstein 1730737
    Synonym Diisopropyl
    Health Hazard 2 GHS H Statement
    H225-H304-H315-H336
    Highly flammable liquid and vapour.
    May be fatal if swallowed and enters airways.
    Causes skin irritation.
    May cause drowsiness or dizziness.
    Flash Point −29°C (−20°F)
    CAS 79-29-8
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P210-P261-P301+P310-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 2,3-DIMETHYLBUTANE
    Boiling Point 57° to 59°C
    Chemical Name or Material 2,3-Dimethylbutane
    Assay 99%

    Description

    Applications
    2,3-Dimethylbutane is a branched saturated hydrocarbon for proteomics research. It is used in organic synthesis, catalytic agent, petrochemical additive.

    Solubility
    Soluble in water (1mg/ml @22°C), DMSO, ethanol, acetone.

    Notes
    Incompatible with strong oxidizing agents, oxygen, ammonia.

    GHS H Statement
    H225-H304-H315-H336
    Highly flammable liquid and vapour.
    May be fatal if swallowed and enters airways.
    Causes skin irritation.
    May cause drowsiness or dizziness.

    GHS P Statement
    P210-P261-P301+P310-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit