Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 2,3-Dimethylbutane, 99%
Click to view available options
Quantity:
25g
100g
Description
Applications
2,3-Dimethylbutane is a branched saturated hydrocarbon for proteomics research. It is used in organic synthesis, catalytic agent, petrochemical additive.
Solubility
Soluble in water (1mg/ml @22°C), DMSO, ethanol, acetone.
Notes
Incompatible with strong oxidizing agents, oxygen, ammonia.
2,3-Dimethylbutane is a branched saturated hydrocarbon for proteomics research. It is used in organic synthesis, catalytic agent, petrochemical additive.
Solubility
Soluble in water (1mg/ml @22°C), DMSO, ethanol, acetone.
Notes
Incompatible with strong oxidizing agents, oxygen, ammonia.
Specifications
Refractive Index | 1.375 |
Density | 0.662 |
Melting Point | -129°C |
Flash Point | −29°C (−20°F) |
Quantity | 25g |
Odor | Mild |
Boiling Point | 57°C to 59°C |
UN Number | UN2457 |
Beilstein | 1730737 |
Solubility Information | Soluble in water (1mg/ml @22°C),DMSO,ethanol,acetone. |
Show More |
Safety and Handling
GHS H Statement
H225-H304-H315-H336
Highly flammable liquid and vapour.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
H225-H304-H315-H319-H336
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 2,3-DIMETHYLBUTANE
EINECSNumber : 201-193-6
RTECSNumber : EJ9350000
TSCA : Yes
Recommended Storage : Ambient temperatures