Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2,3-Dimethylmaleic anhydride, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
50 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS Min % | 96.0 |
CAS Max % | 100.0 |
Melting Point | 91.0°C to 96.0°C |
Boiling Point | 223.0°C |
Assay Percent Range | 97% |
Packaging | Glass bottle |
Quantity | 5 g |
Beilstein | 17, 445 |
Solubility Information | Solubility in water: slightly soluble. Other solubilities: freely soluble in ethanol,ether and chloroform |
Formula Weight | 126.11 |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Warning |
Hazard Statement | GHS H Statement Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Immediately call a POISON CENTER or doctor/physician. IF ON SKIN: Gently wash with plenty of soap and water. IF SWALLOWED: rinse mouth. Do |
EINECSNumber | 212-165-8 |
RTECSNumber | ON4025000 |
TSCA | TSCA |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction