Alfa Aesar™ 2,3-Dimethylphenol, 99%

Manufacturer: Alfa Aesar™ B2286022

Quantity 100g

 View more versions of this product

Catalog No. AAB2286022

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 526-75-0
    Molecular Formula C8H10O
    Formula Weight 122.17
    MDL Number MFCD00002227
    UN Number UN2261
    Beilstein 1906267
    Merck Index 14,10082
    Synonym 2,3-Xylenol
    Chemical Name or Material 2,3-Dimethylphenol

    Specifications

    Formula Weight 122.17
    Synonym 2,3-Xylenol
    RTECS Number ZE5500000
    Boiling Point 218°C
    TSCA TSCA
    Molecular Formula C8H10O
    MDL Number MFCD00002227
    CAS 526-75-0
    UN Number UN2261
    Beilstein 1906267
    Merck Index 14,10082
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H300-H310-H314-H318
    Fatal if swallowed.
    Fatal in contact with skin.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: XYLENOLS, SOLID
    Flash Point 95°C (203°F)
    Melting Point 70° to 73°C
    Quantity 100g
    Chemical Name or Material 2,3-Dimethylphenol
    Assay 99%

    Description

    Applications
    2,3-Dimethylphenol is used for organic synthesis. It is used as the hardener to produce industrial furane, the raw material of producing xylenol specialized product and the assistant of the main raw material of detergent.

    Solubility
    Slightly soluble in water. Soluble in most solvent and aqueous alkali.

    Notes
    Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.

    GHS H Statement
    H300-H310-H314-H318
    Fatal if swallowed.
    Fatal in contact with skin.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit