Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,3-Dimethylphenol, 99%

Manufacturer:  Alfa Aesar™ B2286036

Quantity 500g

 View more versions of this product

Catalog No. AAB2286036


  Check Availability
Add to cart

Specifications

Description

Applications
2,3-Dimethylphenol is used for organic synthesis. It is used as the hardener to produce industrial furane, the raw material of producing xylenol specialized product and the assistant of the main raw material of detergent.

Solubility
Slightly soluble in water. Soluble in most solvent and aqueous alkali.

Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

526-75-0
122.17
UN2261
14,10082
2,3-Dimethylphenol
C8H10O
MFCD00002227
1906267
2,3-Xylenol

Specifications

95°C (203°F)
70° to 73°C
526-75-0
122.17
UN2261
14,10082
2,3-Dimethylphenol
218°C
500g
C8H10O
MFCD00002227
1906267
2,3-Xylenol
99%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H300-H310-H314-H318
Fatal if swallowed.
Fatal in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: XYLENOLS, SOLID

RTECSNumber : ZE5500000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit