Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimethylphenol 99%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Glass bottle
526-75-0
C8H10O
122.167
MFCD00002227
QWBBPBRQALCEIZ-UHFFFAOYSA-N
1-hydroxy-2,3-dimethylbenzene,2,3-xylenol,dimethylphenol,xylenol,phenol, 2,3-dimethyl,o-xylenol,vic-o-xylenol,o-3-xylenol,xyellenol 100,2,3-dimethyl phenol
10687
2,3-dimethylphenol
CC1=C(C(=CC=C1)O)C
526-75-0
C8H10O
122.167
MFCD00002227
QWBBPBRQALCEIZ-UHFFFAOYSA-N
1-hydroxy-2,3-dimethylbenzene,2,3-xylenol,dimethylphenol,xylenol,phenol, 2,3-dimethyl,o-xylenol,vic-o-xylenol,o-3-xylenol,xyellenol 100,2,3-dimethyl phenol
10687
2,3-dimethylphenol
CC1=C(C(=CC=C1)O)C
Melting Point 72°C to 74°C
Quantity 500g
Packaging Glass bottle
Color Brown
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula (CH3)2C6H3OH
Beilstein 06, 480
Merck Index 15, 10279
Formula Weight 122.17
Physical Form Crystalline Solid
Percent Purity ≥98.5%
Chemical Name or Material 2, 3-Dimethylphenol, 99%
Show More Show Less

GHS H Statement:
Toxic to aquatic life with long lasting effects.
Toxic if swallowed.
Toxic in contact with skin.
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 208-395-3

RTECSNumber : ZE5500000

TSCA : TSCA