Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
(2,3-Dimethylphenoxy)acetic acid, 98%, Acros Organics
Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
Specifications
Melting Point | 186°C to 190°C |
Quantity | 1g |
Packaging | Glass bottle |
Assay Percent Range | 97.5% min. (HPLC) |
Linear Formula | (CH3)2C6H3OCH2CO2H |
Solubility Information | Solubility in water: insoluble |
Formula Weight | 180.2 |
Percent Purity | 98% |
Chemical Name or Material | (2, 3-Dimethylphenoxy)acetic acid |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 220-911-9