Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2,3-Dimethylphenoxy)acetic acid, 98%, Acros Organics

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
2935-63-9
C10H12O3
180.2
AVDBMBYECMTQJQ-UHFFFAOYSA-N
2,3-dimethylphenoxyacetic acid,2-2,3-dimethylphenoxy acetic acid,2,3-dimethylphenoxy acetic acid,2,3-xylyloxyacetic acid,acmc-20aog1,enamine_005510,2,3-dimethylphenoxyessig,cambridge id 5161587,2,3-dimethylphenoxy acetic aci,acetic acid, 2,3-dimethylphenoxy
76239
2-(2,3-dimethylphenoxy)acetic acid
CC1=C(C(=CC=C1)OCC(=O)O)C
2935-63-9
C10H12O3
180.2
AVDBMBYECMTQJQ-UHFFFAOYSA-N
2,3-dimethylphenoxyacetic acid,2-2,3-dimethylphenoxy acetic acid,2,3-dimethylphenoxy acetic acid,2,3-xylyloxyacetic acid,acmc-20aog1,enamine_005510,2,3-dimethylphenoxyessig,cambridge id 5161587,2,3-dimethylphenoxy acetic aci,acetic acid, 2,3-dimethylphenoxy
76239
2-(2,3-dimethylphenoxy)acetic acid
CC1=C(C(=CC=C1)OCC(=O)O)C
Melting Point 186°C to 190°C
Quantity 5g
Packaging Glass bottle
Assay Percent Range 97.5% min. (HPLC)
Linear Formula (CH3)2C6H3OCH2CO2H
Solubility Information Solubility in water: insoluble
Formula Weight 180.2
Percent Purity 98%
Chemical Name or Material (2, 3-Dimethylphenoxy)acetic acid

GHS Signal Word: Warning

EINECSNumber : 220-911-9