Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimethylphenyl isocyanate, 99+%, ACROS Organics™

Click to view available options
Quantity:
5g
1591-99-7
C9H9NO
147.177
KNHJIEOCVVIBIV-UHFFFAOYSA-N
2,3-dimethylphenyl isocyanate,2,3-dimethylphenylisocyanate,benzene, isocyanatodimethyl,2,3-dimethylbenzenisocyanate,dimethylphenyl isocyanate,acmc-1bttc,1-isocyanato-2,3-dimethyl-benzene,#,benzene,1-isocyanato-2,3-dimethyl,2,3-dimethylphenyl isocyanate
137096
CHEBI:63899
1-isocyanato-2,3-dimethylbenzene
CC1=C(C(=CC=C1)N=C=O)C
1591-99-7
C9H9NO
147.177
KNHJIEOCVVIBIV-UHFFFAOYSA-N
2,3-dimethylphenyl isocyanate,2,3-dimethylphenylisocyanate,benzene, isocyanatodimethyl,2,3-dimethylbenzenisocyanate,dimethylphenyl isocyanate,acmc-1bttc,1-isocyanato-2,3-dimethyl-benzene,#,benzene,1-isocyanato-2,3-dimethyl,2,3-dimethylphenyl isocyanate
137096
CHEBI:63899
1-isocyanato-2,3-dimethylbenzene
CC1=C(C(=CC=C1)N=C=O)C
Refractive Index 1.5380 to 1.5400 (20°C, 589nm)
Quantity 5g
Color Yellow
Infrared Spectrum Authentic
Density 1.059g/mL
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Linear Formula (CH3)C6H3NCO
Formula Weight 147.18
Physical Form Liquid
Percent Purity ≥99.0%
Chemical Name or Material 2, 3-Dimethylphenyl isocyanate
Show More Show Less

Recommended Storage : Refrigerator +4°C