Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,3-Dimethylquinoxaline, 97%

Click to view available options
Quantity:
5g
25g
100g
2379-55-7
C10H10N2
158.204
MFCD00006728
FKHNZQFCDGOQGV-UHFFFAOYSA-N
quinoxaline, 2,3-dimethyl,aurora ka-3449,quinoxaline,3-dimethyl,2,3-dimethyl-quinoxalin,2,3-dimethyl-quinoxaline,acmc-1cg5s,quinoxaline,2,3-dimethyl,salor-int l498777-1ea,5-23-07-00267 beilstein handbook reference,chembl67865
16925
2,3-dimethylquinoxaline
CC1=NC2=CC=CC=C2N=C1C
2379-55-7
C10H10N2
158.204
MFCD00006728
FKHNZQFCDGOQGV-UHFFFAOYSA-N
quinoxaline, 2,3-dimethyl,aurora ka-3449,quinoxaline,3-dimethyl,2,3-dimethyl-quinoxalin,2,3-dimethyl-quinoxaline,acmc-1cg5s,quinoxaline,2,3-dimethyl,salor-int l498777-1ea,5-23-07-00267 beilstein handbook reference,chembl67865
16925
2,3-dimethylquinoxaline
CC1=NC2=CC=CC=C2N=C1C
Quantity 25g
Melting Point 102°C to 107°C
Beilstein 114832
Formula Weight 158.2
Percent Purity 97%
Chemical Name or Material 2,3-Dimethylquinoxaline

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : VD1960000

TSCA : TSCA